missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RP3-366L4.2 Partial ORF (NP_741999.3, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00150274-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102127

  • 335.00€ / 10µg

Estimated Shipment: 09-05-2023
to see stock.
Add to basket

Description

Description

Sequence: MWRGRAGALLRVWGFWPTGVPRRRPLSCDAASQAGSNYPRCWNCGGPWGPGREDRFFCPQCRALQAPDPTRDYFSLMDCNRSFRVDTAKLQHRYQQLQRLVHPDFFSQRS
Specifications

Specifications

NP_741999.3
Liquid
150274
RP3-366L4.2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DNAJC20/HSC20/JAC1/MGC2637/MGC74462/dJ366L4.2
HSCB
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MWRGRAGALLRVWGFWPTGVPRRRPLSCDAASQAGSNYPRCWNCGGPWGPGREDRFFCPQCRALQAPDPTRDYFSLMDCNRSFRVDTAKLQHRYQQLQRLVHPDFFSQRS
RUO
HSCB
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers