Learn More
Abnova™ Human RNF41 Partial ORF (NP_005776, 218 a.a. - 317 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010193-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. The specific function of this protein has not yet been determined. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported. [provided by RefSeq]
Sequence: ISTPDAVLQAVIKRSLVESGCPASIVNELIENAHERSWPQGLATLETRQMNRRYYENYVAKRIPGKQAVVVMACENQHMGDDMVQEPGLVMIFAHGVEEISpecifications
NP_005776 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ISTPDAVLQAVIKRSLVESGCPASIVNELIENAHERSWPQGLATLETRQMNRRYYENYVAKRIPGKQAVVVMACENQHMGDDMVQEPGLVMIFAHGVEEI | |
RUO | |
RNF41 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10193 | |
RNF41 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC45228/NRDP1/SBBI03 | |
RNF41 | |
Recombinant | |
wheat germ expression system |