missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF39 Partial ORF (NP_079512.1, 321 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_079512.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80352 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100307
|
Abnova™
H00080352-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16190297
|
Abnova™
H00080352-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: QRKGCVRLCPAGAVWAVEGRGGRLWALTAPEPTLLGGVEPPPRRIRVDLDWERGRVAFYDGRSLDLLYAFQAPGPLGERIFPLFCTCDPRAPLRIVPAESSpecifications
NP_079512.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HZF/HZFW/LIRF | |
RNF39 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
80352 | |
RNF39 (Human) Recombinant Protein (Q01) | |
QRKGCVRLCPAGAVWAVEGRGGRLWALTAPEPTLLGGVEPPPRRIRVDLDWERGRVAFYDGRSLDLLYAFQAPGPLGERIFPLFCTCDPRAPLRIVPAES | |
RUO | |
RNF39 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |