missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF20 Partial ORF (NP_062538, 177 a.a. - 276 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056254-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192163

  • 349.00€ / 10µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene shares similarity with BRE1 of S. cerevisiae. Yeast BRE1 is a ubiquitin ligase required for the ubiquitination of histone H2B and the methylation of histone H3. [provided by RefSeq]

Sequence: RVESSRRAVSQIVTVYDKLQEKVELLSRKLNSGDNLIVEEAVQELNSFLAQENMRLQELTDLLQEKHRTMSQEFSKLQSKVETAESRVSVLESMIDDLQW
Specifications

Specifications

NP_062538
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RVESSRRAVSQIVTVYDKLQEKVELLSRKLNSGDNLIVEEAVQELNSFLAQENMRLQELTDLLQEKHRTMSQEFSKLQSKVETAESRVSVLESMIDDLQW
RUO
RNF20
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
56254
RNF20 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BRE1/BRE1A/FLJ11189/FLJ20382/KIAA2779/MGC129667/MGC129668/hBRE1
RNF20
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers