Learn More
Abnova™ Human RNF139 Full-length ORF (NP_009149.2, 1 a.a. - 664 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011236-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a multi-membrane spanning protein containing a RING-H2 finger. This protein is located in the endoplasmic reticulum, and has been shown to possess ubiquitin ligase activity. This gene was found to be interrupted by a t(3:8) translocation in a family with hereditary renal and non-medulary thyroid cancer. Studies of the Drosophila counterpart suggested that this protein may interact with tumor suppressor protein VHL, as well as with COPS5/JAB1, a protein responsible for the degradation of tumor suppressor CDKN1B/P27KIP. [provided by RefSeq]
Sequence: MAAVGPPQQQVRMAHQQVWAALEVALRVPCLYIIDAIFNSYPDSSQSRFCIVLQIFLRLFGVFASSIVLILSQRSLFKFYTYSSAFLLAATSVLVNYYASLHIDFYGAYNTSAFGIELLPRKGPSLWMALIVLQLTFGIGYVTLLQIHSIYSQLIILDLLVPVIGLITELPLHIRETLLFTSSLILTLNTVFVLAVKLKWFYYSTRYVYLLVRHMYRIYGLQLLMEDTWKRIRFPDILRVFWLTRVTAQATVLMYILRMANETDSFFISWDDFWDLICNLIISGCDSTLTVLGMSAVISSVAHYLGLGILAFIGSTEEDDRRLGFVAPVLFFILALQTGLSGLRPEERLIRLSRNMCLLLTAVLHFIHGMTDPVLMSLSASHVSSFRRHFPVLFVSACLFILPVLLSYVLWHHYALNTWLFAVTAFCVELCLKVIVSLTVYTLFMIDGYYNVLWEKLDDYVYYVRSTGSIIEFIFGVVMFGNGAYTMMFESGSKIRAFMMCLHAYFNIYLQAKNGWKTFMNRRTAVKKINSLPEIKGSRLQEINDVCAICYHEFTTSARITPCNHYFHALCLRKWLYIQDTCPMCHQKVYIEDDIKDNSNVSNNNGFIPPNETPEEAVREAAAESDRELNEDDSTDCDDDVQRERNGVIQHTGAAAEEFNDDTDSpecifications
NP_009149.2 | |
Liquid | |
11236 | |
RNF139 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAVGPPQQQVRMAHQQVWAALEVALRVPCLYIIDAIFNSYPDSSQSRFCIVLQIFLRLFGVFASSIVLILSQRSLFKFYTYSSAFLLAATSVLVNYYASLHIDFYGAYNTSAFGIELLPRKGPSLWMALIVLQLTFGIGYVTLLQIHSIYSQLIILDLLVPVIGLITELPLHIRETLLFTSSLILTLNTVFVLAVKLKWFYYSTRYVYLLVRHMYRIYGLQLLMEDTWKRIRFPDILRVFWLTRVTAQATVLMYILRMANETDSFFISWDDFWDLICNLIISGCDSTLTVLGMSAVISSVAHYLGLGILAFIGSTEEDDRRLGFVAPVLFFILALQTGLSGLRPEERLIRLSRNMCLLLTAVLHFIHGMTDPVLMSLSASHVSSFRRHFPVLFVSACLFILPVLLSYVLWHHYALNTWLFAVTAFCVELCLKVIVSLTVYTLFMIDGYYNVLWEKLDDYVYYVRSTGSIIEFIFGVVMFGNGAYTMMFESGSKIRAFMMCLHAYFNIYLQAKNGWKTFMNRRTAVKKINSLPEIKGSRLQEINDVCAICYHEFTTSARITPCNHYFHALCLRKWLYIQDTCPMCHQKVYIEDDIKDNSNVSNNNGFIPPNETPEEAVREAAAESDRELNEDDSTDCDDDVQRERNGVIQHTGAAAEEFNDDTD | |
RUO | |
RNF139 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
102.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HRCA1/MGC31961/RCA1/TRC8 | |
RNF139 | |
Yes | |
wheat germ expression system |