missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF12 Partial ORF (NP_057204, 1 a.a. - 83 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051132-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16166626

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a RING-H2 zinc finger protein. It has been shown to be an E3 ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MENSDSNDKGSGDQSAAQRRSQMDRLDREEAFYQFVNNLSEEDYRLMRDNNLLGTPGESTEEELLRRLQQIKEGPPPQNSDEN
Specifications

Specifications

NP_057204
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.87kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MENSDSNDKGSGDQSAAQRRSQMDRLDREEAFYQFVNNLSEEDYRLMRDNNLLGTPGESTEEELLRRLQQIKEGPPPQNSDEN
RUO
RNF12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51132
RNF12 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC15161/NY-REN-43/RLIM
RNF12
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers