Learn More
R&D Systems™ Human RNase3 (NP_002926.2) VersaClone cDNA
Specifications
Specifications
| Gene | RNASE3 |
| Accession Number | NP_002926.2 |
| Gene ID (Entrez) | 6037 |
| Gene Alias | eosinophil cationic protein, ECP, ribonuclease, RNase A family, 3, RNS3 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |