missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RHOXF1 Partial ORF (NP_644811.1, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00158800-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MARSLVHDTVFYCLSVYQVKISPTPQLGAASSAEGHVGQGAPGLMGNMNPEGGVNHENGMNRDGGMIPEGGGGNQEPRQQPQPPPEEPAQAAMEGPQPENMQPRTRRTKFSpecifications
NP_644811.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MARSLVHDTVFYCLSVYQVKISPTPQLGAASSAEGHVGQGAPGLMGNMNPEGGVNHENGMNRDGGMIPEGGGGNQEPRQQPQPPPEEPAQAAMEGPQPENMQPRTRRTKF | |
RUO | |
RHOXF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
158800 | |
RHOXF1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119030/MGC119033/OTEX/PEPP1 | |
RHOXF1 | |
Recombinant | |
wheat germ expression system |