missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RHBDL2 Full-length ORF (AAH13103.1, 1 a.a. - 121 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054933-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190923

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. The encoded protein is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3. [provided by RefSeq]

Sequence: MNLNMGREMKEELEEEEKMREDGGGKDRAKSKKVHRIVSKWMLPEKSRGTYLERANCFPPPVFIISISLAELAVFIYYAVWKPQKQWITLDTGILESPFIYSPEKREEAWRFISYMLVHAG
Specifications

Specifications

AAH13103.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
40.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC16997/RRP2
RHBDL2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54933
RHBDL2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNLNMGREMKEELEEEEKMREDGGGKDRAKSKKVHRIVSKWMLPEKSRGTYLERANCFPPPVFIISISLAELAVFIYYAVWKPQKQWITLDTGILESPFIYSPEKREEAWRFISYMLVHAG
RUO
RHBDL2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers