Learn More
Abnova™ Human RGS2 Full-length ORF (NP_002914.1, 1 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005997-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 2 belongs to this family. The protein acts as a mediator of myeloid differentiation and may play a role in leukemogenesis. [provided by RefSeq]
Sequence: MQSAMFLAVQHDCRPMDKSAGSGHKSEEKREKMKRTLLKDWKTRLSYFLQNSSTPGKPKTGKKSKQQAFIKPSPEEAQLWSEAFDELLASKYGLAAFRAFLKSEFCEENIEFWLACEDFKKTKSPQKLSSKARKIYTDFIEKEAPKEINIDFQTKTLIAQNIQEATSGCFTTAQKRVYSLMENNSYPRFLESEFYQDLCKKPQITTEPHATSpecifications
NP_002914.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
G0S8 | |
RGS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5997 | |
RGS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQSAMFLAVQHDCRPMDKSAGSGHKSEEKREKMKRTLLKDWKTRLSYFLQNSSTPGKPKTGKKSKQQAFIKPSPEEAQLWSEAFDELLASKYGLAAFRAFLKSEFCEENIEFWLACEDFKKTKSPQKLSSKARKIYTDFIEKEAPKEINIDFQTKTLIAQNIQEATSGCFTTAQKRVYSLMENNSYPRFLESEFYQDLCKKPQITTEPHAT | |
RUO | |
RGS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |