missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RGS17 Partial ORF (NP_036551, 111 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026575-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195546

  • 528.00€ / 25µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of the regulator of G-protein signaling family. This protein contains a conserved, 120 amino acid motif called the RGS domain and a cysteine-rich region. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. [provided by RefSeq]

Sequence: LLFWLACEDLKKEQNKKVIEEKARMIYEDYISILSPKEVSLDSRVREVINRNLLDPNPHMYEDAQLQIYTLMHRDSFPRFLNSQIYKSFVESTAGSSSES
Specifications

Specifications

NP_036551
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LLFWLACEDLKKEQNKKVIEEKARMIYEDYISILSPKEVSLDSRVREVINRNLLDPNPHMYEDAQLQIYTLMHRDSFPRFLNSQIYKSFVESTAGSSSES
RUO
RGS17
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26575
RGS17 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RGS-17/RGSZ2/hRGS17
RGS17
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers