missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RGL2 Partial ORF (AAH32681, 644 a.a. - 743 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005863-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: PGASDCRIIRVQMELGEDGSVYKSILVTSQDKAPSVISRVLKKNNRDSAVASEYELVQLLPGERELTIPASANVFYAMDGASHDFLLRQRRRSSTATPGVSpecifications
AAH32681 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PGASDCRIIRVQMELGEDGSVYKSILVTSQDKAPSVISRVLKKNNRDSAVASEYELVQLLPGERELTIPASANVFYAMDGASHDFLLRQRRRSSTATPGV | |
RUO | |
RGL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5863 | |
RGL2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HKE1.5/KE1.5/RAB2L | |
RGL2 | |
Recombinant | |
wheat germ expression system |