Learn More
Abnova™ Human RGC32 Partial ORF (NP_054778.1, 1 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_054778.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 28984 |
Molecular Weight (g/mol) | 38.61kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025455
|
Abnova™
H00028984-Q02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16015455
|
Abnova™
H00028984-Q02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene is thought to regulate cell cycle progression. It is induced by p53 in response to DNA damage, or by sublytic levels of complement system proteins that result in activation of the cell cycle. The encoded protein localizes to the cytoplasm during interphase and to centrosomes during mitosis. The protein forms a complex with polo-like kinase 1. The protein also translocates to the nucleus in response to treatment with complement system proteins, and can associate with and increase the kinase activity of cell division cycle 2 protein. In different assays and cell types, overexpression of this protein has been shown to activate or suppress cell cycle progression. [provided by RefSeq]
Sequence: MKPPAEDLSDALCEFDAVLADFASPFHERHFHYEEHLERMKRRSSASVSDSSGFSDSESADSLYRNSFSFSDEKLNSPTDSTPALLSATVTPQKAKLGDTKELEAFIADLDKTLASMSpecifications
NP_054778.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.61kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0564/MGC87338/RGC-32/RGC32/bA157L14.2 | |
C13orf15 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
28984 | |
RGC32 (Human) Recombinant Protein (Q02) | |
MKPPAEDLSDALCEFDAVLADFASPFHERHFHYEEHLERMKRRSSASVSDSSGFSDSESADSLYRNSFSFSDEKLNSPTDSTPALLSATVTPQKAKLGDTKELEAFIADLDKTLASM | |
RUO | |
C13orf15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |