Learn More
Abnova™ Human RFXANK Full-length ORF (NP_604389.1, 1 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008625-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. This protein contains ankyrin repeats involved in protein-protein interactions. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group B. Two transcript variants encoding different isoforms have been described for this gene, with only one isoform showing activation activity. [provided by RefSeq]
Sequence: MELTQPAEDLIQTQQTPASELGDPEDPGEEAADGSDTVVLSLFPCTPEPVNPEPDASVSSPQGSSLKHSTTLTNRQRGNEVSALPATLDCDNLVNKPDERGFTPLIWASAFGEIETVRFLLEWGADPHILAKERESALSLASTGGYTDIVGLLLERDVDINIYDWNGGTPLLYAVRGNHVKCVEALLARGADLTTEADSGYTPMDLAVALGYRKVQQVIENHILKLFQSNLVPADPESpecifications
NP_604389.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANKRA1/BLS/F14150_1/MGC138628/RFX-B | |
RFXANK | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8625 | |
RFXANK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MELTQPAEDLIQTQQTPASELGDPEDPGEEAADGSDTVVLSLFPCTPEPVNPEPDASVSSPQGSSLKHSTTLTNRQRGNEVSALPATLDCDNLVNKPDERGFTPLIWASAFGEIETVRFLLEWGADPHILAKERESALSLASTGGYTDIVGLLLERDVDINIYDWNGGTPLLYAVRGNHVKCVEALLARGADLTTEADSGYTPMDLAVALGYRKVQQVIENHILKLFQSNLVPADPE | |
RUO | |
RFXANK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |