missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Resistin Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Gene ID (Entrez) | 56729 |
| Synonym | Adipose tissue-specific secretory factor, ADSF, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN, RETN1, RSTNXCP1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction