missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human REPS2 Partial ORF (NP_004717, 66 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009185-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120116

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The product of this gene is part of a protein complex that regulates the endocytosis of growth factor receptors. The encoded protein directly interacts with a GTPase activating protein that functions downstream of the small G protein Ral. Its expression can negatively affect receptor internalization and inhibit growth factor signaling. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: RVPLSHGYSKLRSSAEQMHPAPYEARQPLVQPEGSSSGGPGTKPLRHQASLIRSFSVERELQDNSSYPDEPWRITEEQREYYVNQFRSLQPDPSSFISGS
Specifications

Specifications

NP_004717
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RVPLSHGYSKLRSSAEQMHPAPYEARQPLVQPEGSSSGGPGTKPLRHQASLIRSFSVERELQDNSSYPDEPWRITEEQREYYVNQFRSLQPDPSSFISGS
RUO
REPS2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9185
REPS2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
POB1
REPS2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers