missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RCBTB2 Partial ORF (NP_001259, 86 a.a. - 194 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001102-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106254

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the RCC1-related GEF family. The N-terminal half of the encoded amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran. [provided by RefSeq]

Sequence: TIEPRRLDSLNGKKIACLSYGSGPHIVLATTEGEVFTWGHNAYSQLGNGTTNHGLVPCHISTNLSNKQVIEVACGSYHSLVLTSDGEVFAWGYNNSGQVGSGSTVNQPI
Specifications

Specifications

NP_001259
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TIEPRRLDSLNGKKIACLSYGSGPHIVLATTEGEVFTWGHNAYSQLGNGTTNHGLVPCHISTNLSNKQVIEVACGSYHSLVLTSDGEVFAWGYNNSGQVGSGSTVNQPI
RUO
RCBTB2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1102
RCBTB2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CHC1L
RCBTB2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers