Learn More
Abnova™ Human RBP2 Full-length ORF (NP_004155.2, 1 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005948-P01.10ug
Additional Details : Weight : 0.00010kg
Description
RBP2 is an abundant protein present in the small intestinal epithelium. It is thought to participate in the uptake and/or intracellular metabolism of vitamin A. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. RBP2 may also modulate the supply of retinoic acid to the nuclei of endometrial cells during the menstrual cycle. [provided by RefSeq]
Sequence: MTRDQNGTWEMESNENFEGYMKALDIDFATRKIAVRLTQTKVIDQDGDNFKTKTTSTFRNYDVDFTVGVEFDEYTKSLDNRHVKALVTWEGDVLVCVQKGEKENRGWKQWIEGDKLYLELTCGDQVCRQVFKKKSpecifications
NP_004155.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CRABP-II/CRBP2/CRBPII/RBPC2 | |
RBP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5948 | |
RBP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTRDQNGTWEMESNENFEGYMKALDIDFATRKIAVRLTQTKVIDQDGDNFKTKTTSTFRNYDVDFTVGVEFDEYTKSLDNRHVKALVTWEGDVLVCVQKGEKENRGWKQWIEGDKLYLELTCGDQVCRQVFKKK | |
RUO | |
RBP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |