missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RBBP5 Partial ORF (NP_005048.2, 392 a.a. - 491 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005048.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5929 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124165
|
Abnova™
H00005929-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment:
Log in to see stock. |
|||||
16134165
|
Abnova™
H00005929-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment:
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that bind directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. [provided by RefSeq]
Sequence: ELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSKSpecifications
NP_005048.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RBQ3/SWD1 | |
RBBP5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5929 | |
RBBP5 (Human) Recombinant Protein (Q01) | |
ELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSK | |
RUO | |
RBBP5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |