Learn More
Abnova™ Human RBBP5 Full-length ORF (NP_005048.2, 1 a.a. - 538 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005929-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that bind directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. [provided by RefSeq]
Sequence: MNLELLESFGQNYPEEADGTLDCISMALTCTFNRWGTLLAVGCNDGRIVIWDFLTRGIAKIISAHIHPVCSLCWSRDGHKLVSASTDNIVSQWDVLSGDCDQRFRFPSPILKVQYHPRDQNKVLVCPMKSAPVMLTLSDSKHVVLPVDDDSDLNVVASFDRRGEYIYTGNAKGKILVLKTDSQDLVASFRVTTGTSNTTAIKSIEFARKGSCFLINTADRIIRVYDGREILTCGRDGEPEPMQKLQDLVNRTPWKKCCFSGDGEYIVAGSARQHALYIWEKSIGNLVKILHGTRGELLLDVAWHPVRPIIASISSGVVSIWAQNQVENWSAFAPDFKELDENVEYEERESEFDIEDEDKSEPEQTGADAAEDEEVDVTSVDPIAAFCSSDEELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSKGKEKDSPFKPKLYKGDRGLPLEGSAKGKVQAELSQPLTAGGAISELLSpecifications
NP_005048.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
85.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RBQ3/SWD1 | |
RBBP5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5929 | |
RBBP5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNLELLESFGQNYPEEADGTLDCISMALTCTFNRWGTLLAVGCNDGRIVIWDFLTRGIAKIISAHIHPVCSLCWSRDGHKLVSASTDNIVSQWDVLSGDCDQRFRFPSPILKVQYHPRDQNKVLVCPMKSAPVMLTLSDSKHVVLPVDDDSDLNVVASFDRRGEYIYTGNAKGKILVLKTDSQDLVASFRVTTGTSNTTAIKSIEFARKGSCFLINTADRIIRVYDGREILTCGRDGEPEPMQKLQDLVNRTPWKKCCFSGDGEYIVAGSARQHALYIWEKSIGNLVKILHGTRGELLLDVAWHPVRPIIASISSGVVSIWAQNQVENWSAFAPDFKELDENVEYEERESEFDIEDEDKSEPEQTGADAAEDEEVDVTSVDPIAAFCSSDEELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSKGKEKDSPFKPKLYKGDRGLPLEGSAKGKVQAELSQPLTAGGAISELL | |
RUO | |
RBBP5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |