missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RASSF2 Full-length ORF (NP_055552.1, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009770-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104102

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein that contains a Ras association domain. Similar to its cattle and sheep counterparts, this gene is located near the prion gene. Two alternatively spliced transcripts encoding the same isoform have been reported. [provided by RefSeq]

Sequence: MDYSHQTSLVPCGQDKYISKNELLLHLKTYNLYYEGQNLQLRHREEEDEFIVEGLLNISWGLRRPIRLQMQDDNERIRPPPSSSSWHSGCNLGAQGTTLKPLTVPKVQISEVDAPPEGDQMPSSTDSRGLKPLQEDTPQLMRTRSDVGVRRRGNVRTPSDQRRIRRHRFSINGHFYNHKTSVFTPAYGSVTNVRINSTMTTPQVLKLLLNKFKIENSAEEFALYVVHTSGEKQKLKATDYPLIARILQGPCEQISKVFLMEKDQVEEVTYDVAQYIKFEMPVLKSFIQKLQEEEDREVKKLMRKYTVLRLMIRQRLEEIAETPATI
Specifications

Specifications

NP_055552.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781O1747/KIAA0168/RASFADIN
RASSF2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9770
RASSF2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDYSHQTSLVPCGQDKYISKNELLLHLKTYNLYYEGQNLQLRHREEEDEFIVEGLLNISWGLRRPIRLQMQDDNERIRPPPSSSSWHSGCNLGAQGTTLKPLTVPKVQISEVDAPPEGDQMPSSTDSRGLKPLQEDTPQLMRTRSDVGVRRRGNVRTPSDQRRIRRHRFSINGHFYNHKTSVFTPAYGSVTNVRINSTMTTPQVLKLLLNKFKIENSAEEFALYVVHTSGEKQKLKATDYPLIARILQGPCEQISKVFLMEKDQVEEVTYDVAQYIKFEMPVLKSFIQKLQEEEDREVKKLMRKYTVLRLMIRQRLEEIAETPATI
RUO
RASSF2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers