missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RASD1 Partial ORF (AAH18041, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051655-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157196

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a Ras-related protein that is stimulated by dexamethasone. The exact function of this gene is unknown, but it may play a role in dexamethasone-induced alterations in cell morphology, growth and cell-extracellular matrix interactions. In addition, studies of a similar rat protein suggest that it functions as as a novel physiologic nitric oxide (NO) effector. The gene product belongs to the Ras superfamily of small GTPases. [provided by RefSeq]

Sequence: MKLAAMIKKMCPSDSELSIPAKNCYRMVILGSSKVGKTAIVSRFLTGRFEDAYTPTIEDFHRKFYSIRGEVYQLDILDTSGNHPFPAMRRLSILTGDVFILVFSLDNRDS
Specifications

Specifications

AAH18041
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKLAAMIKKMCPSDSELSIPAKNCYRMVILGSSKVGKTAIVSRFLTGRFEDAYTPTIEDFHRKFYSIRGEVYQLDILDTSGNHPFPAMRRLSILTGDVFILVFSLDNRDS
RUO
RASD1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51655
RASD1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AGS1/DEXRAS1/MGC:26290
RASD1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers