missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAET1G Partial ORF (NP_001001788.1, 124 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00353091-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192877

  • 528.00€ / 25µg

Estimated Shipment: 03-11-2023
to see stock.
Add to basket

Description

Description

Members of the RAET1 family, such as RAET1G, are major histocompatibility complex (MHC) class I-related genes located within a 180-kb cluster on chromosome 6q24.2-q25.3. RAET1 proteins contain MHC class I-like alpha-1 and alpha-2 domains. RAET1E (MIM 609243) and RAET1G differ from the other RAET1 proteins (e.g., RAET1I, or ULBP1; MIM 605697) in that they have type I membrane-spanning sequences at their C termini rather than glycosylphosphatidylinositol anchor sequences. (Radosavljevic et al., 2002 [PubMed 11827464]).[supplied by OMIM]

Sequence: RMSCEQKAEGHGSGSWQLSFDGQIFLLFDSENRMWTTVHPGARKMKEKWENDKDMTMSFHYISMGDCTGWLEDFLMGMDSTLEPSAGGTV
Specifications

Specifications

NP_001001788.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RMSCEQKAEGHGSGSWQLSFDGQIFLLFDSENRMWTTVHPGARKMKEKWENDKDMTMSFHYISMGDCTGWLEDFLMGMDSTLEPSAGGTV
RUO
RAET1G
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
353091
RAET1G (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RAET1G
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers