missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAD54B Partial ORF (AAH01965, 801 a.a. - 910 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00025788-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157732

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene belongs to the DEAD-like helicase superfamily. It shares similarity with Saccharomyces cerevisiae RAD54 and RDH54, both of which are involved in homologous recombination and repair of DNA. This protein binds to double-stranded DNA, and displays ATPase activity in the presence of DNA. This gene is highly expressed in testis and spleen, which suggests active roles in meiotic and mitotic recombination. Homozygous mutations of this gene were observed in primary lymphoma and colon cancer. [provided by RefSeq]

Sequence: HIQFSVEELKNLFTLHESSDCVTHDLLDCECTGEEVHTGDSLEKFIVSRDCQLGPHHQKSNSLKPLSMSQLKQWKHFSGDHLNLTDPFLERITENVSFIFQNITTQATGT
Specifications

Specifications

AAH01965
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HIQFSVEELKNLFTLHESSDCVTHDLLDCECTGEEVHTGDSLEKFIVSRDCQLGPHHQKSNSLKPLSMSQLKQWKHFSGDHLNLTDPFLERITENVSFIFQNITTQATGT
RUO
RAD54B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
25788
RAD54B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FSBP/RDH54
RAD54B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers