Learn More
Abcam Human RAB3A knockout SK-N-FI cell line
Product Code. 32153897 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32153897 1,000,000 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32153897 Supplier Abcam Supplier No. ab2887081000000Cells_
 Restricted Item
Estimated Shipment: 16-11-2026
RAB3A KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Homozygote. 62 bp deletion. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select 1000000 Cells/vial.
RAB3A KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Homozygote. 62 bp deletion. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells