missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAB34 Partial ORF (NP_114140.2, 170 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083871-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100557

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

RAB proteins, like RAB34, are small GTPases that regulate vesicle budding, docking, and fusion along endocytosis and exocytosis pathways (Chen et al., 2003 [PubMed 12684051]).[supplied by OMIM]

Sequence: LSTPAQYALMEKDALQVAQEMKAEYWAVSSLTGENVREFFFRVAALTFEANVLAELEKSGARRIGDVVRINSDDSNLYLTASKKKPTCCP
Specifications

Specifications

NP_114140.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LSTPAQYALMEKDALQVAQEMKAEYWAVSSLTGENVREFFFRVAALTFEANVLAELEKSGARRIGDVVRINSDDSNLYLTASKKKPTCCP
RUO
RAB34
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
83871
RAB34 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RAB39/RAH
RAB34
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers