Learn More
Abnova™ Human RAB28 Full-length ORF (AAH18067, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009364-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the Rab subfamily of Ras-related small GTPases. The encoded protein may be involved in regulating intracellular trafficking. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 9 and X. [provided by RefSeq]
Sequence: MSDSEEESQDRQLKIVVLGDGASGKTSLTTCFAQETFGKQYKQTIGLDFFLRRITLPGNLNVTLQIWDIGGQTIGGKMLDKYIYGAQLIWSICEQSpecifications
AAH18067 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC41862 | |
RAB28 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9364 | |
RAB28 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSDSEEESQDRQLKIVVLGDGASGKTSLTTCFAQETFGKQYKQTIGLDFFLRRITLPGNLNVTLQIWDIGGQTIGGKMLDKYIYGAQLIWSICEQ | |
RUO | |
RAB28 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |