Learn More
Human RAB1B knockout HEK-293T cell lysate, 1 Kit
Product Code. 32143061 Shop All Abcam Products
Product Code. 32143061 Supplier Abcam Supplier No. ab2590741Kit
 Restricted Item
Estimated Shipment: 10-11-2026
Human RAB1B knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: RAB1B knockout HEK-293T