missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PYGO1 Full-length ORF (AAI46573.1, 1 a.a. - 419 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026108-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MPAENSPAPAYKVSSHGGDSGLDGLGGPGVQLGSPDKKKRKANTQGPSFPPLSEYAPPPNPNSDHLVAANPFDDNYNTISYKPLPSSNPYLGPGYPGFGGYSTFRMPPHVPPRMSSPYCGPYSLRNQPHPFPQNPLGMGFNRPHAFNFGPHDNSSFGNPSYNNALSQNVNMPNQHFRQNPAENFSQIPPQNASQVSNPDLASNFVPGNNSNFTSPLESNHSFIPPPNTFGQAKAPPPKQDFTQGATKNTNQNSSAHPPHLNMDDTVNQSNIELKNVNRNNAVNQENSRSSSTEATNNNPANGTQNKPRQPRGAADACTTEKSNKSSLHPNRHGHSSSDPVYPCGICTNEVNDDQDAILCEASCQKWFHRICTGMTETAYGLLTAEASAVWGCDTCMADKDVQLMRTRETFGPSAVGSDASpecifications
AAI46573.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.04kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp547G0910 | |
PYGO1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26108 | |
PYGO1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPAENSPAPAYKVSSHGGDSGLDGLGGPGVQLGSPDKKKRKANTQGPSFPPLSEYAPPPNPNSDHLVAANPFDDNYNTISYKPLPSSNPYLGPGYPGFGGYSTFRMPPHVPPRMSSPYCGPYSLRNQPHPFPQNPLGMGFNRPHAFNFGPHDNSSFGNPSYNNALSQNVNMPNQHFRQNPAENFSQIPPQNASQVSNPDLASNFVPGNNSNFTSPLESNHSFIPPPNTFGQAKAPPPKQDFTQGATKNTNQNSSAHPPHLNMDDTVNQSNIELKNVNRNNAVNQENSRSSSTEATNNNPANGTQNKPRQPRGAADACTTEKSNKSSLHPNRHGHSSSDPVYPCGICTNEVNDDQDAILCEASCQKWFHRICTGMTETAYGLLTAEASAVWGCDTCMADKDVQLMRTRETFGPSAVGSDA | |
RUO | |
PYGO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |