Learn More
Abnova™ Human PXDN Partial ORF (XP_056455, 1452 a.a. - 1561 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007837-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.[supplied by OMIM]
Sequence: STSAFSTRSDASGTNDFREFVLEMQKTITDLRTQIKKLESRLSTTECVDAGGESHANNTKWKKDACTICECKDGQVTCFVEACPPATCAVPVNIPGACCPVCLQKRAEEKSpecifications
XP_056455 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
STSAFSTRSDASGTNDFREFVLEMQKTITDLRTQIKKLESRLSTTECVDAGGESHANNTKWKKDACTICECKDGQVTCFVEACPPATCAVPVNIPGACCPVCLQKRAEEK | |
RUO | |
PXDN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7837 | |
PXDN (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D2S448/D2S448E/KIAA0230/MG50/PRG2/PXN/VPO | |
PXDN | |
Recombinant | |
wheat germ expression system |