missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PURA Partial ORF (NP_005850.1, 92 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005813-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16094635

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia. [provided by RefSeq]

Sequence: GAGGNKSRLTLSMSVAVEFRDYLGDFIEHYAQLGPSQPPDLAQAQDEPRR
Specifications

Specifications

NP_005850.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
30.91kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GAGGNKSRLTLSMSVAVEFRDYLGDFIEHYAQLGPSQPPDLAQAQDEPRR
RUO
PURA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5813
PURA (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PUR-ALPHA/PUR1/PURALPHA
PURA
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers