Learn More
Abnova™ Human PTPRJ Partial ORF (NP_002834.2, 38 a.a. - 137 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005795-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes, including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region containing five fibronectin type III repeats, a single transmembrane region, and a single intracytoplasmic catalytic domain, and thus represents a receptor-type PTP. This protein is present in all hematopoietic lineages, and was shown to negatively regulate T cell receptor signaling possibly through interfering with the phosphorylation of Phospholipase C Gamma 1 and Linker for Activation of T Cells. This protein can also dephosphorylate the PDGF beta receptor, and may be involved in UV-induced signal transduction. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GTPSPIPDPSVATVATGENGITQISSTAESFHKQNGTGTPQVETNTSEDGESSGANDSLRTPEQGSNGTDGASQKTPSSTGPSPVFDIKAVSISPTNVILSpecifications
NP_002834.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GTPSPIPDPSVATVATGENGITQISSTAESFHKQNGTGTPQVETNTSEDGESSGANDSLRTPEQGSNGTDGASQKTPSSTGPSPVFDIKAVSISPTNVIL | |
RUO | |
PTPRJ | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5795 | |
PTPRJ (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD148/DEP1/HPTPeta/R-PTP-ETA/SCC1 | |
PTPRJ | |
Recombinant | |
wheat germ expression system |