missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTPN4 Full-length ORF (NP_002821.1, 1 a.a. - 926 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005775-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16159891

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. This PTP has been shown to interact with glutamate receptor delta 2 and epsilon subunits, and is thought to play a role in signalling downstream of the glutamate receptors through tyrosine dephosphorylation. [provided by RefSeq]

Sequence: MTSRFRLPAGRTYNVRASELARDRQHTEVVCNILLLDNTVQAFKVNKHDQGQVLLDVVFKHLDLTEQDYFGLQLADDSTDNPRWLDPNKPIRKQLKRGSPYSLNFRVKFFVSDPNKLQEEYTRYQYFLQIKQDILTGRLPCPSNTAALLASFAVQSELGDYDQSENLSGYLSDYSFIPNQPQDFEKEIAKLHQQHIGLSPAEAEFNYLNTARTLELYGVEFHYARDQSNNEIMIGVMSGGILIYKNRVRMNTFPWLKIVKISFKCKQFFIQLRKELHESRETLLGFNMVNYRACKNLWKACVEHHTFFRLDRPLPPQKNFFAHYFTLGSKFRYCGRTEVQSVQYGKEKANKDRVFARSPSKPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDGKPPALPPKQSKKNSWNQIHYSHSQQDLESHINETFDIPSSPEKPTPNGGIPHDNLVLIRMKPDENGRFGFNVKGGYDQKMPVIVSRVAPGTPADLCVPRLNEGDQVVLINGRDIAEHTHDQVVLFIKASCERHSGELMLLVRPNAVYDVVEEKLENEPDFQYIPEKAPLDSVHQDDHSLRESMIQLAEGLITGTVLTQFDQLYRKKPGMTMSCAKLPQNISKNRYRDISPYDATRVILKGNEDYINANYINMEIPSSSIINQYIACQGPLPHTCTDFWQMTWEQGSSMVVMLTTQVERGRVKCHQYWPEPTGSSSYGCYQVTCHSEEGNTAYIFRKMTLFNQEKNESRPLTQIQYIAWPDHGVPDDSSDFLDFVCHVRNKRAGKEEPVVVHCSAGIGRTGVLITMETAMCLIECNQPVYPLDIVRTMRDQRAMMIQTPSQYRFVCEAILKVYEEGFVKPLTTSTNK
Specifications

Specifications

NP_002821.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
132.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PTPMEG/PTPMEG1
PTPN4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5775
PTPN4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSRFRLPAGRTYNVRASELARDRQHTEVVCNILLLDNTVQAFKVNKHDQGQVLLDVVFKHLDLTEQDYFGLQLADDSTDNPRWLDPNKPIRKQLKRGSPYSLNFRVKFFVSDPNKLQEEYTRYQYFLQIKQDILTGRLPCPSNTAALLASFAVQSELGDYDQSENLSGYLSDYSFIPNQPQDFEKEIAKLHQQHIGLSPAEAEFNYLNTARTLELYGVEFHYARDQSNNEIMIGVMSGGILIYKNRVRMNTFPWLKIVKISFKCKQFFIQLRKELHESRETLLGFNMVNYRACKNLWKACVEHHTFFRLDRPLPPQKNFFAHYFTLGSKFRYCGRTEVQSVQYGKEKANKDRVFARSPSKPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDGKPPALPPKQSKKNSWNQIHYSHSQQDLESHINETFDIPSSPEKPTPNGGIPHDNLVLIRMKPDENGRFGFNVKGGYDQKMPVIVSRVAPGTPADLCVPRLNEGDQVVLINGRDIAEHTHDQVVLFIKASCERHSGELMLLVRPNAVYDVVEEKLENEPDFQYIPEKAPLDSVHQDDHSLRESMIQLAEGLITGTVLTQFDQLYRKKPGMTMSCAKLPQNISKNRYRDISPYDATRVILKGNEDYINANYINMEIPSSSIINQYIACQGPLPHTCTDFWQMTWEQGSSMVVMLTTQVERGRVKCHQYWPEPTGSSSYGCYQVTCHSEEGNTAYIFRKMTLFNQEKNESRPLTQIQYIAWPDHGVPDDSSDFLDFVCHVRNKRAGKEEPVVVHCSAGIGRTGVLITMETAMCLIECNQPVYPLDIVRTMRDQRAMMIQTPSQYRFVCEAILKVYEEGFVKPLTTSTNK
RUO
PTPN4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers