missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTK9 Partial ORF (AAH22344, 80 a.a. - 252 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005756-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193795

  • 349.00€ / 10µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1. [provided by RefSeq]

Sequence: VAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSSAKPKGPAGKRGIRRLIRGPAETEATTD
Specifications

Specifications

AAH22344
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.66kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSSAKPKGPAGKRGIRRLIRGPAETEATTD
RUO
TWF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5756
PTK9 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A6/MGC23788/MGC41876/PTK9
TWF1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers