Learn More
ACROBiosystems Human PTH1R / PTHR1 Protein, His Tag
Brand: ACROBiosystems PTR-H5222-1MG
Additional Details : Weight : 0.00001kg
Description
Parathyroid hormone/parathyroid hormone-related peptide receptor is also known as Parathyroid hormone 1 receptor (PTH1R), PTHR1, PTH/PTHrP type I receptor (PTH/PTHr receptor), which belongs to the G-protein coupled receptor 2 family. PTH1R / PTHR1 is a receptor for parathyroid hormone and for parathyroid hormone-related peptide. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.6 kDa. The protein migrates as 27-43 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Specifications
NP_000307.1 | |
1 mg | |
Asp 27 - Gly 188 | |
PTH1R,PTHR,PTHR1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |
19.6 kDa | |
Human PTH1R, His Tag (PTR-H5222) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Gly 188 (Accession # Q03431-1). | |
-20°C | |
PTH1R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |