missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PSMD12 Full-length ORF (NP_002807.1, 1 a.a. - 456 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005718-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101928

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator. A pseudogene has been identified on chromosome 3. [provided by RefSeq]

Sequence: MADGGSERADGRIVKMEVDYSATVDQRLPECAKLAKEGRLQEVIETLLSLEKQTRTASDMVSTSRILVAVVKMCYEAKEWDLLNENIMLLSKRRSQLKQAVAKMVQQCCTYVEEITDLPIKLRLIDTLRMVTEGKIYVEIERARLTKTLATIKEQNGDVKEAASILQELQVETYGSMEKKERVEFILEQMRLCLAVKDYIRTQIISKKINTKFFQEENTEKLKLKYYNLMIQLDQHEGSYLSICKHYRAIYDTPCIQAESEKWQQALKSVVLYVILAPFDNEQSDLVHRISGDKKLEEIPKYKDLLKLFTTMELMRWSTLVEDYGMELRKGSLESPATDVFGSTEEGEKRWKDLKNRVVEHNIRIMAKYYTRITMKRMAQLLDLSVDESEAFLSNLVVNKTIFAKVDRLAGIINFQRPKDPNNLLNDWSQKLNSLMSLVNKTTHLIAKEEMIHNLQ
Specifications

Specifications

NP_002807.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
79.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC75406/Rpn5/p55
PSMD12
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5718
PSMD12 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADGGSERADGRIVKMEVDYSATVDQRLPECAKLAKEGRLQEVIETLLSLEKQTRTASDMVSTSRILVAVVKMCYEAKEWDLLNENIMLLSKRRSQLKQAVAKMVQQCCTYVEEITDLPIKLRLIDTLRMVTEGKIYVEIERARLTKTLATIKEQNGDVKEAASILQELQVETYGSMEKKERVEFILEQMRLCLAVKDYIRTQIISKKINTKFFQEENTEKLKLKYYNLMIQLDQHEGSYLSICKHYRAIYDTPCIQAESEKWQQALKSVVLYVILAPFDNEQSDLVHRISGDKKLEEIPKYKDLLKLFTTMELMRWSTLVEDYGMELRKGSLESPATDVFGSTEEGEKRWKDLKNRVVEHNIRIMAKYYTRITMKRMAQLLDLSVDESEAFLSNLVVNKTIFAKVDRLAGIINFQRPKDPNNLLNDWSQKLNSLMSLVNKTTHLIAKEEMIHNLQ
RUO
PSMD12
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers