Learn More
R&D Systems™ Human Pro-Collagen I alpha 1 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Specifications
Specifications
| Assay Range | 78.10 - 5000 pg/mL |
| Assay Sensitivity | 3.51 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Plasma (Heparin), Saliva, Serum, Tissue Lysates, Urine |
| Specificity | Natural and recombinant human Pro-Collagen I alpha 1. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1277 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | Alpha-1 type I collagen, COL1A1, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain |
| Target | Human |