missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRG1 Partial ORF (NP_002718, 29 a.a. - 94 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005552-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193265

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

This gene encodes a protein best known as a hematopoietic cell granule proteoglycan. Proteoglycans stored in the secretory granules of many hematopoietic cells also contain a protease-resistant peptide core, which may be important for neutralizing hydrolytic enzymes. This encoded protein was found to be associated with the macromolecular complex of granzymes and perforin, which may serve as a mediator of granule-mediated apoptosis. [provided by RefSeq]

Sequence: PTRRARYQWVRCNPDSNSANCLEEKGPMFELLPGESNKIPRLRTDLFPKTRIQDLNRIFPLSEDYS
Specifications

Specifications

NP_002718
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PTRRARYQWVRCNPDSNSANCLEEKGPMFELLPGESNKIPRLRTDLFPKTRIQDLNRIFPLSEDYS
RUO
SRGN
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5552
PRG1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12930/MGC9289/PPG/PRG/PRG1
SRGN
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers