Learn More
Abnova™ Human PRDM2 Partial ORF (NP_036363, 1606 a.a. - 1705 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007799-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This tumor suppressor gene is a member of a nuclear histone/protein methyltransferase superfamily. It encodes a zinc finger protein that can bind to retinoblastoma protein, estrogen receptor, and the TPA-responsive element (MTE) of the heme-oxygenase-1 gene. Although the functions of this protein have not been fully characterized, it may (1) play a role in transcriptional regulation during neuronal differentiation and pathogenesis of retinoblastoma, (2) act as a transcriptional activator of the heme-oxygenase-1 gene, and (3) be a specific effector of estrogen action. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: KTSRSLHVRVQKSKAVLQSKSTLASKKRTDRFNIKSRERSGGPVTRSLQLAAAADLSENKREDGSAKQELKDFSYSLRLASRCSPPAAPYITRQYRKVKASpecifications
NP_036363 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KTSRSLHVRVQKSKAVLQSKSTLASKKRTDRFNIKSRERSGGPVTRSLQLAAAADLSENKREDGSAKQELKDFSYSLRLASRCSPPAAPYITRQYRKVKA | |
RUO | |
PRDM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7799 | |
PRDM2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUMHOXY1/KMT8/MTB-ZF/RIZ/RIZ1/RIZ2 | |
PRDM2 | |
Recombinant | |
wheat germ expression system |