Learn More
Abnova™ Human PPT1 Full-length ORF (NP_000301.1, 1 a.a. - 306 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005538-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene
Sequence: MASPGCLWLLAVALLPWTCASRALQHLDPPAPLPLVIWHGMGDSCCNPLSMGAIKKMVEKKIPGIYVLSLEIGKTLMEDVENSFFLNVNSQVTTVCQALAKDPKLQQGYNAMGFSQGGQFLRAVAQRCPSPPMINLISVGGQHQGVFGLPRCPGESSHICDFIRKTLNAGAYSKVVQERLVQAEYWHDPIKEDVYRNHSIFLADINQERGINESYKKNLMALKKFVMVKFLNDSIVDPVDSEWFGFYRSGQAKETIPLQETSLYTQDRLGLKEMDNAGQLVFLATEGDHLQLSEEWFYAHIIPFLGSpecifications
NP_000301.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLN1/INCL/PPT | |
PPT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5538 | |
PPT1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASPGCLWLLAVALLPWTCASRALQHLDPPAPLPLVIWHGMGDSCCNPLSMGAIKKMVEKKIPGIYVLSLEIGKTLMEDVENSFFLNVNSQVTTVCQALAKDPKLQQGYNAMGFSQGGQFLRAVAQRCPSPPMINLISVGGQHQGVFGLPRCPGESSHICDFIRKTLNAGAYSKVVQERLVQAEYWHDPIKEDVYRNHSIFLADINQERGINESYKKNLMALKKFVMVKFLNDSIVDPVDSEWFGFYRSGQAKETIPLQETSLYTQDRLGLKEMDNAGQLVFLATEGDHLQLSEEWFYAHIIPFLG | |
RUO | |
PPT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |