missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPM1F Full-length ORF (NP_055449.1, 1 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009647-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193962

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase can interact with Rho guanine nucleotide exchange factors (PIX), and thus block the effects of p21-activated kinase 1 (PAK), a protein kinase mediating biological effects downstream of Rho GTPases. Calcium/calmodulin-dependent protein kinase II gamma (CAMK2G/CAMK-II) is found to be one of the substrates of this phosphatase. The overexpression of this phosphatase or CAMK2G has been shown to mediate caspase-dependent apoptosis. An alternatively spliced transcript variant has been identified, but its full-length nature has not been determined. [provided by RefSeq]

Sequence: MSSGAPQKSSPMASGAEETPGFLDTLLQDFPALLNPEDPLPWKAPGTVLSQEEVEGELAELAMGFLGSRKAPPPLAAALAHEAVSQLLQTDLSEFRKLPREEEEEEEDDDEEEKAPVTLLDAQSLAQSFFNRLWEVAGQWQKQVPLAARASQRQWLVSIHAIRNTRRKMEDRHVSLPSFNQLFGLSDPVNRAYFAVFDGHGGVDAARYAAVHVHTNAARQPELPTDPEGALREAFRRTDQMFLRKAKRERLQSGTTGVCALIAGATLHVAWLGDSQVILVQQGQVVKLMEPHRPERQDEKARIEALGGFVSHMDCWRVNGTLAVSRAIGDVFQKPYVSGEADAASRALTGSEDYLLLACDGFFDVVPHQEVVGLVQSHLTRQQGSGLRVAEELVAAARERGSHDNITVMVVFLRDPQELLEGGNQGEGDPQAEGRRQDLPSSLPEPETQAPPRS
Specifications

Specifications

NP_055449.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CaMKPase/FEM-2/KIAA0015/POPX2/hFEM-2
PPM1F
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9647
PPM1F (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSGAPQKSSPMASGAEETPGFLDTLLQDFPALLNPEDPLPWKAPGTVLSQEEVEGELAELAMGFLGSRKAPPPLAAALAHEAVSQLLQTDLSEFRKLPREEEEEEEDDDEEEKAPVTLLDAQSLAQSFFNRLWEVAGQWQKQVPLAARASQRQWLVSIHAIRNTRRKMEDRHVSLPSFNQLFGLSDPVNRAYFAVFDGHGGVDAARYAAVHVHTNAARQPELPTDPEGALREAFRRTDQMFLRKAKRERLQSGTTGVCALIAGATLHVAWLGDSQVILVQQGQVVKLMEPHRPERQDEKARIEALGGFVSHMDCWRVNGTLAVSRAIGDVFQKPYVSGEADAASRALTGSEDYLLLACDGFFDVVPHQEVVGLVQSHLTRQQGSGLRVAEELVAAARERGSHDNITVMVVFLRDPQELLEGGNQGEGDPQAEGRRQDLPSSLPEPETQAPPRS
RUO
PPM1F
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers