missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPIL4 Full-length ORF (NP_624311.1, 1 a.a. - 492 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00085313-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16116283

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. [provided by RefSeq]

Sequence: MAVLLETTLGDVVIDLYTEERPRACLNFLKLCKIKYYNYCLIHNVQRDFIIQTGDPTGTGRGGESIFGQLYGDQASFFEAEKVPRIKHKKKGTVSMVNNGSDQHGSQFLITTGENLDYLDGVHTVFGEVTEGMDIIKKINETFVDKDFVPYQDIRINHTVILDDPFDDPPDLLIPDRSPEPTREQLDSGRIGADEEIDDFKGRSAEEVEEIKAEKEAKTQAILLEMVGDLPDADIKPPENVLFVCKLNPVTTDEDLEIIFSRFGPIRSCEVIRDWKTGESLCYAFIEFEKEEDCEKAFFKMDNVLIDDRRIHVDFSQSVAKVKWKGKGGKYTKSDFKEYEKEQDKPPNLVLKDKVKPKQDTKYDLILDEQAEDSKSSHSHTSKKHKKKTHHCSEEKEDEDYMPIKNTNQDIYREMGFGHYEEEESCWEKQKSEKRDRTQNRSRSRSRERDGHYSNSHKSKYQTDLYERERSKKRDRSRSPKKSKDKEKSKYR
Specifications

Specifications

NP_624311.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
83.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HDCME13P
PPIL4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
85313
PPIL4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVLLETTLGDVVIDLYTEERPRACLNFLKLCKIKYYNYCLIHNVQRDFIIQTGDPTGTGRGGESIFGQLYGDQASFFEAEKVPRIKHKKKGTVSMVNNGSDQHGSQFLITTGENLDYLDGVHTVFGEVTEGMDIIKKINETFVDKDFVPYQDIRINHTVILDDPFDDPPDLLIPDRSPEPTREQLDSGRIGADEEIDDFKGRSAEEVEEIKAEKEAKTQAILLEMVGDLPDADIKPPENVLFVCKLNPVTTDEDLEIIFSRFGPIRSCEVIRDWKTGESLCYAFIEFEKEEDCEKAFFKMDNVLIDDRRIHVDFSQSVAKVKWKGKGGKYTKSDFKEYEKEQDKPPNLVLKDKVKPKQDTKYDLILDEQAEDSKSSHSHTSKKHKKKTHHCSEEKEDEDYMPIKNTNQDIYREMGFGHYEEEESCWEKQKSEKRDRTQNRSRSRSRERDGHYSNSHKSKYQTDLYERERSKKRDRSRSPKKSKDKEKSKYR
RUO
PPIL4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers