missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPIE Partial ORF (NP_006103, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010450-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192536

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein contains a highly conserved cyclophilin (CYP) domain as well as an RNA-binding domain. It was shown to possess PPIase and protein folding activities and also exhibit RNA-binding activity. Three alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: ATTKRVLYVGGLAEEVDDKVLHAAFIPFGDITDIQIPLDYETEKHRGFAFVEFELAEDAAAAIDNMNESELFGRTIRVNLAKPMRIKEGSSRPVWSDDD
Specifications

Specifications

NP_006103
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ATTKRVLYVGGLAEEVDDKVLHAAFIPFGDITDIQIPLDYETEKHRGFAFVEFELAEDAAAAIDNMNESELFGRTIRVNLAKPMRIKEGSSRPVWSDDD
RUO
PPIE
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10450
PPIE (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CYP-33/MGC111222/MGC3736
PPIE
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers