Learn More
Abnova™ Human PPCDC Full-length ORF (AAH14409.1, 1 a.a. - 204 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00060490-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCDC (EC 4.1.1.36), one of the last enzymes in this pathway, converts phosphopantothenoylcysteine to 4-prime-phosphopantetheine (Daugherty et al., 2002 [PubMed 11923312]).[supplied by OMIM]
Sequence: MEPKASCPAAAPLMERKFHVLVGVTGSVAALKLPLLVSKLLDIPGLEVAVVTTERAKHFYSPQDIPVTLYSDADEWEMWKSRSDPVLHIDLRRWADLLLVAPLDANTLGKVASGICDNLLTCVMRAWDRSKPLLFCPAMNTAMWEHPITAQQVDQLKAFGYVEIPCVAKKLVCGDEGLGAMAEVGTIVDKVKEVLFQHSGFQQSSpecifications
AAH14409.1 | |
Liquid | |
60490 | |
PPCDC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPKASCPAAAPLMERKFHVLVGVTGSVAALKLPLLVSKLLDIPGLEVAVVTTERAKHFYSPQDIPVTLYSDADEWEMWKSRSDPVLHIDLRRWADLLLVAPLDANTLGKVASGICDNLLTCVMRAWDRSKPLLFCPAMNTAMWEHPITAQQVDQLKAFGYVEIPCVAKKLVCGDEGLGAMAEVGTIVDKVKEVLFQHSGFQQS | |
RUO | |
PPCDC | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14585/MDS018 | |
PPCDC | |
Yes | |
wheat germ expression system |