Learn More
Abnova™ Human POFUT2 Full-length ORF (NP_598368.2, 1 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023275-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor (EGF; MIM 131530)-like repeats or thrombospondin (THBS; see MIM 188060) type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates (Luo et al., 2006 [PubMed 16464857]).[supplied by OMIM]
Sequence: MATLSFVFLLLGAVSWPPASASGQEFWPGQSAADILSGAASRRRYLLYDVNPPEGFNLRRDVYIRIASLLKTLLKTEEWVLVLPPWGRLYHWQSPDIHQVRIPWSEFFDLPSLNKNIPVIEYEQFIAESGGPFIDQVYVLQSYAEGWKEGTWEEKVDERPCIDQLLYSQDKHEYYRGWFWGYEETRGLNVSCLSVQGSASIVAPLLLRNTSARSVMLDRAENLLHDHYGGKEYWDTRRSMVFARHLREVGDEFRSRHLNSTDDADRIPFQEDWMKMKVKLGSALGGPYLGVHLRRKDFIWGHRQDVPSLEGAVRKIRSLMKTHRLDKVFVATDAVRKEYEELKKLLPEMVRFEPTWEELELYKDGGVAIIDQWICAHARFFIGTSVSTFSFRIHEEREILGLDPKTTYNRFCGDQEKACEQPTHWKITYSpecifications
NP_598368.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C21orf80/FUT13 | |
POFUT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23275 | |
POFUT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATLSFVFLLLGAVSWPPASASGQEFWPGQSAADILSGAASRRRYLLYDVNPPEGFNLRRDVYIRIASLLKTLLKTEEWVLVLPPWGRLYHWQSPDIHQVRIPWSEFFDLPSLNKNIPVIEYEQFIAESGGPFIDQVYVLQSYAEGWKEGTWEEKVDERPCIDQLLYSQDKHEYYRGWFWGYEETRGLNVSCLSVQGSASIVAPLLLRNTSARSVMLDRAENLLHDHYGGKEYWDTRRSMVFARHLREVGDEFRSRHLNSTDDADRIPFQEDWMKMKVKLGSALGGPYLGVHLRRKDFIWGHRQDVPSLEGAVRKIRSLMKTHRLDKVFVATDAVRKEYEELKKLLPEMVRFEPTWEELELYKDGGVAIIDQWICAHARFFIGTSVSTFSFRIHEEREILGLDPKTTYNRFCGDQEKACEQPTHWKITY | |
RUO | |
POFUT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |