Learn More
R&D Systems™ Human Podocin/NPHS2 (NP_055440.1) VersaClone cDNA
Specifications
Specifications
| Gene | Podocin/NPHS2 |
| Accession Number | NP_055440.1 |
| Concentration | 10 μg (at 0.2 μg/μL) |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1165 bp |
| Gene ID (Entrez) | 7827 |
| Gene Alias | Nephrosis 2, nephrosis 2, idiopathic, steroid-resistant (podocin), NPHS2, PDCN, PDCNSRN1podocin, SRN1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |