Learn More
Abcam Human PLEKHA4 knockout A549 cell line
Product Code. 32140800
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32140800 1,000,000 Cells Each
32140491 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32140800 Supplier Abcam Supplier No. ab2669771000000Cells_
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PLEKHA4 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 6 and 2 bp deletion in exon 6. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knock...
PLEKHA4 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 6 and 2 bp deletion in exon 6. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells