missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLA2G3 Partial ORF (NP_056530, 21 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00050487-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106376

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

PLA2G3 belongs to the family of secreted phospholipase A2 (sPLA2; EC 3.1.1.4) proteins. These Ca(2+)-dependent lipolytic enzymes have a conserved Ca(2+)-binding loop and a his-asp dyad in the catalytic site (Murakami et al., 2003 [PubMed 12522102]).[supplied by OMIM]

Sequence: SPALRWYRTSCHLTKAVPGNPLGYLSFLAKDAQGLALIHARWDAHRRLQACSWEDEPELTAAYGALCAHETAWGSFIHTPGPELQRALATLQSQWEACRALEESPAGARK
Specifications

Specifications

NP_056530
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SPALRWYRTSCHLTKAVPGNPLGYLSFLAKDAQGLALIHARWDAHRRLQACSWEDEPELTAAYGALCAHETAWGSFIHTPGPELQRALATLQSQWEACRALEESPAGARK
RUO
PLA2G3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
50487
PLA2G3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GIII-SPLA2/SPLA2III
PLA2G3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers