missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLA1A Partial ORF (NP_056984, 241 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051365-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106906

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Phosphatidylserine-specific phospholipase A1-alpha (PLA1A) acts specifically on phosphatidylserine (PS) and 1-acyl-2-lysophosphatidylserine (lyso-PS) to hydrolyze fatty acids at the sn-1 position of these phospholipids.[supplied by OMIM]

Sequence: DQPGCPTFFYAGYSYLICDHMRAVHLYISALENSCPLMAFPCASYKAFLAGRCLDCFNPFLLSCPRIGLVEQGGVKIEPLPKEVKVYLLTTSSAPYCMHH
Specifications

Specifications

NP_056984
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DQPGCPTFFYAGYSYLICDHMRAVHLYISALENSCPLMAFPCASYKAFLAGRCLDCFNPFLLSCPRIGLVEQGGVKIEPLPKEVKVYLLTTSSAPYCMHH
RUO
PLA1A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51365
PLA1A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PS-PLA1/PSPLA1
PLA1A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers