Learn More
Abnova™ Human PKIG Partial ORF (NP_861521, 1 a.a. - 76 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011142-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the cAMP-dependent protein kinase (PKA) inhibitor family. Studies of a similar protein in mice suggest that this protein acts as a potent competitive PKA inhibitor, and is a predominant form of PKA inhibitors in various tissues. Three alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]
Sequence: MMEVESSYSDFISCDRTGRRNAVPDIQGDSEAVSVRKLAGDMGELALEGAEGQVEGSAPDKEAGNQPQSSDGTTSSSpecifications
NP_861521 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMEVESSYSDFISCDRTGRRNAVPDIQGDSEAVSVRKLAGDMGELALEGAEGQVEGSAPDKEAGNQPQSSDGTTSS | |
RUO | |
PKIG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11142 | |
PKIG (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126458/MGC126459 | |
PKIG | |
Recombinant | |
wheat germ expression system |